missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-N-isopropyl-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59205.03

Code : NEW

Additional Details : CAS Number : 349096-38-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405448

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

N-Benzyl-N-isopropyl-3-methylbenzamide
349096-38-4
267.37
MFCD00784147

Specifications

N-Benzyl-N-isopropyl-3-methylbenzamide
Liquid
267.37
Pale yellow
349096-38-4
97%
MFCD00784147
1g
Videos
SDS