missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-N-isopropyl-3,5-dinitrobenzamide, 97%

Brand:  Alfa Aesar™ H59345.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405628

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyl-N-isopropyl-3,5-dinitrobenzamide
343.33
MFCD00783877

Specifications

N-Benzyl-N-isopropyl-3,5-dinitrobenzamide
97%
MFCD00783877
Solid
343.33
1g
Videos
SDS