missing translation for 'onlineSavingsMsg'
Learn More

N-(Benzyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog Number p-7037041
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
837-18-3
C13H13NO2S
247.312
MFCD00092709
GRTPAOVVVLZLDP-UHFFFAOYSA-N
benzenesulfonamide, n-benzyl, n-benzyl-benzenesulfonamide, benzenesulfonamide, n-phenylmethyl, n-benzyl benzenesulfonamide, maybridge1_001915, n-phenylsulfonylbenzylamine, benzyl phenylsulfonyl amine, 2-12-00-00571 beilstein handbook reference
95236
N-benzylbenzenesulfonamide
C1=CC=C(C=C1)CNS(=O)(=O)C2=CC=CC=C2
This item is not returnable. View return policy
837-18-3
C13H13NO2S
247.312
MFCD00092709
GRTPAOVVVLZLDP-UHFFFAOYSA-N
benzenesulfonamide, n-benzyl, n-benzyl-benzenesulfonamide, benzenesulfonamide, n-phenylmethyl, n-benzyl benzenesulfonamide, maybridge1_001915, n-phenylsulfonylbenzylamine, benzyl phenylsulfonyl amine, 2-12-00-00571 beilstein handbook reference
95236
N-benzylbenzenesulfonamide
C1=CC=C(C=C1)CNS(=O)(=O)C2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is applied in chemical research and as an active pharmaceutical ingredient.

Notes
Keep away from oxidizing agents and strong acids. Store at room temperature.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 247.31
Percent Purity 97%
Chemical Name or Material N-(Benzyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'