missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-4-chloro-N-isopropylbenzamide, 97%

Brand:  Alfa Aesar™ H59106.03

Code : NEW

Additional Details : CAS Number : 57409-41-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15425318

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyl-4-chloro-N-isopropylbenzamide
57409-41-3
287.79
MFCD00426537

Specifications

N-Benzyl-4-chloro-N-isopropylbenzamide
Solid
287.79
1g
57409-41-3
97%
MFCD00426537
Videos
SDS