missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-4-chloro-3-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65555.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462569

  • £74.73 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyl-4-chloro-3-sulfamoylbenzamide
324.78
MFCD10684235

Specifications

N-Benzyl-4-chloro-3-sulfamoylbenzamide
95%
MFCD10684235
Solid
324.78
1g
Videos
SDS