missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzyl-4-bromo-2-fluoro-N-isopropylbenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H65282.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429536

  • £79.71 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzyl-4-bromo-2-fluoro-N-isopropylbenzenesulfonamide
386.28
MFCD28009654

Specifications

N-Benzyl-4-bromo-2-fluoro-N-isopropylbenzenesulfonamide
97%
MFCD28009654
Solid
386.28
1g
Videos
SDS