missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2-chloro-N-ethyl-4-nitrobenzamide, 97%, Thermo Scientific™

Catalog Number p-7040083
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
349093-97-6
C16H15ClN2O3
318.757
MFCD01212352
XXEMVWJWUFWUQQ-UHFFFAOYSA-N
2-chloro-4-nitrophenyl-n-ethyl-n-benzylcarboxamide
4011077
N-benzyl-2-chloro-N-ethyl-4-nitrobenzamide
CCN(CC1=CC=CC=C1)C(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl
This item is not returnable. View return policy
349093-97-6
C16H15ClN2O3
318.757
MFCD01212352
XXEMVWJWUFWUQQ-UHFFFAOYSA-N
2-chloro-4-nitrophenyl-n-ethyl-n-benzylcarboxamide
4011077
N-benzyl-2-chloro-N-ethyl-4-nitrobenzamide
CCN(CC1=CC=CC=C1)C(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 318.75
Percent Purity 97%
Chemical Name or Material N-Benzyl-2-chloro-N-ethyl-4-nitrobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'