missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2,6-difluoro-N-phenylbenzamide, 97%, Thermo Scientific™

Catalog Number p-7040215
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1426315-84-5
C20H15F2NO
323.343
MFCD22683100
GPQFSRTYCAFMCY-UHFFFAOYSA-N
98000094
N-benzyl-2,6-difluoro-N-phenylbenzamide
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=C(C=CC=C3F)F
This item is not returnable. View return policy
1426315-84-5
C20H15F2NO
323.343
MFCD22683100
GPQFSRTYCAFMCY-UHFFFAOYSA-N
98000094
N-benzyl-2,6-difluoro-N-phenylbenzamide
C1=CC=C(C=C1)CN(C2=CC=CC=C2)C(=O)C3=C(C=CC=C3F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 323.34
Percent Purity 97%
Chemical Name or Material N-Benzyl-2,6-difluoro-N-phenylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'