missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Benzoyl-L-tyrosine ethyl ester, 98%

Brand:  Alfa Aesar™ H63434.14

Code : NEW

Additional Details : CAS Number : 3483-82-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15460129

  • £74.00 / 25g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

N-Benzoyl-L-tyrosine ethyl ester
313.35
2223819
3483-82-7
MFCD00002388
Bz-Tyr-OEt; (S)-Ethyl 2-benzamido-3-(4-hydroxyphenyl)propionate

Specifications

N-Benzoyl-L-tyrosine ethyl ester
Solid
313.35
2223819
118° to 121°C
3483-82-7
98%
MFCD00002388
Bz-Tyr-OEt; (S)-Ethyl 2-benzamido-3-(4-hydroxyphenyl)propionate
25g
Videos
SDS