missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoyl-L-glutamic acid, 98%, Thermo Scientific™

Catalog Number 15422829
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
6094-36-6
C12H13NO5
251.238
MFCD00014028
LPJXPACOXRZCCP-VIFPVBQESA-N
bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid
11345724
(2S)-2-benzamidopentanedioic acid
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O
This item is not returnable. View return policy
6094-36-6
C12H13NO5
251.238
MFCD00014028
LPJXPACOXRZCCP-VIFPVBQESA-N
bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid
11345724
(2S)-2-benzamidopentanedioic acid
C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Beilstein 2622989
Formula Weight 251.24
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Benzoyl-L-glutamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'