missing translation for 'onlineSavingsMsg'
Learn More

N-Arachidonylglycine, Tocris Bioscience™

GPR18 agonist; endocannabinoid

Brand:  Tocris (Bio-Techne) 14455

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 5mg

 View more versions of this product

Product Code. 13449513

  • £130.24 / 5mg

Estimated Shipment
10-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

GPR18 agonist (EC50 = 44.5 nM). Endogenous anandamide-like compound. Lacks affinity for CB1 receptors (Ki > 10 μM), VR1 receptors (EC50 > 10 μM) and anandamide transporters (IC50 > 50 μM) but causes hot-plate analgesia in mice when given orally, and suppresses tonic inflammatory pain. Also endogenous GlyT2 inhibitor.
Specifications

Chemical Identifiers

179113-91-8
361.526
n-arachidonylglycine, n-arachidonoylglycine, nagly, n-arachidonoyl glycine, arachidonoyl glycine, n-5z,8z,11z,14z-eicosatetraenoyl-glycine, dsstox_cid_25124, dsstox_rid_80688, dsstox_gsid_45124, 2-5z,8z,11z,14z-icosa-5,8,11,14-tetraenoyl amino acetic acid
CHEBI:58961
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCC(=O)O
C22H35NO3
YLEARPUNMCCKMP-DOFZRALJSA-N
5283389
2-[[(5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoyl]amino]acetic acid

Specifications

N-Arachidonylglycine
C22H35NO3
YLEARPUNMCCKMP-DOFZRALJSA-N
2-[[(5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoyl]amino]acetic acid
5283389
361.52
5mg
179113-91-8
n-arachidonylglycine, n-arachidonoylglycine, nagly, n-arachidonoyl glycine, arachidonoyl glycine, n-5z,8z,11z,14z-eicosatetraenoyl-glycine, dsstox_cid_25124, dsstox_rid_80688, dsstox_gsid_45124, 2-5z,8z,11z,14z-icosa-5,8,11,14-tetraenoyl amino acetic acid
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCC(=O)O
361.526
CHEBI:58961
>98%
Videos
Documents