missing translation for 'onlineSavingsMsg'
Learn More

n-Amyl acetate, 99%, Thermo Scientific Chemicals

Catalog Number p-3598415
26.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
628-63-7
C7H14O2
130.19
MFCD00009500
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C
This item is not returnable. View return policy
628-63-7
C7H14O2
130.19
MFCD00009500
PGMYKACGEOXYJE-UHFFFAOYSA-N
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether
12348
CHEBI:87362
pentyl acetate
CCCCCOC(=O)C

n-Amyl acetate, 99%, C7H14O2, CAS Number-628-63-7, 1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate, 2.5L, 98.5% min. (GC), 02, 131, 149.0 deg.C, 100.0, CHEBI:87362

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.8°C
Color Colorless
Density 0.8700g/mL
Boiling Point 149.0°C
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2(CH2)4CH3
Refractive Index 1.4010 to 1.4030
Quantity 250 mL
Beilstein 02, 131
Specific Gravity 0.87
Solubility Information Solubility in water: 10g/L (20c). Other solubilities: miscible with alcohol and ether
Formula Weight 130.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Amyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Amyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'