missing translation for 'onlineSavingsMsg'
Learn More

N-Allylaniline, 95%, Thermo Scientific Chemicals

Catalog Number p-7052787
35.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
589-09-3
C9H11N
133.19
MFCD00008638
LQFLWKPCQITJIH-UHFFFAOYSA-N
n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
68525
C=CCNC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
589-09-3
C9H11N
133.19
MFCD00008638
LQFLWKPCQITJIH-UHFFFAOYSA-N
n-allylaniline, n-allyl-aniline, benzenamine, n-2-propenyl, unii-q7s639gwxg, n-prop-2-en-1-ylaniline, q7s639gwxg, phenylprop-2-enylamine, nyl, allylphenylamine, allyl-phenyl-amine
68525
C=CCNC1=CC=CC=C1

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep away from oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Color Colorless to Brown
Density 0.982 g/mL
Boiling Point 218°C to 220°C
Flash Point 89°C (192°F)
Linear Formula C6H5NHCH2CH=CH2
Refractive Index 1.563
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name N-prop-2-enylaniline
Formula Weight 133.19
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-Allylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'