missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Allylaniline, 95%, Thermo Scientific Chemicals
35.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep away from oxidizing agents and strong acids.
It is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep away from oxidizing agents and strong acids.
Specifications
Color | Colorless to Brown |
Density | 0.982 g/mL |
Boiling Point | 218°C to 220°C |
Flash Point | 89°C (192°F) |
Linear Formula | C6H5NHCH2CH=CH2 |
Refractive Index | 1.563 |
Quantity | 5 g |
Solubility Information | Insoluble in water. |
IUPAC Name | N-prop-2-enylaniline |
Formula Weight | 133.19 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'