Learn More
N-Acetyltryptamine, Tocris Bioscience™
Substrate for serotonin N-acetyl transferase
Brand:  Tocris (Bio-Techne) 035750
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13447583
  • £468.93 / 50mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
A partial agonist for melatonin receptors in the retina. Also used for determination of serotonin N-acetyl transferase.
Specifications
Chemical Identifiers
1016-47-3
202.257
n-acetyltryptamine, n-2-1h-indol-3-yl ethyl acetamide, 3-2-n-acetylaminoethyl indole, acetamide, n-2-1h-indol-3-yl ethyl, chembl33171, n-2-1h-indol-3-yl-ethyl-acetamide, acetotryptamide, acetamide, n-2-indol-3-ylethyl
CHEBI:55515
CC(=O)NCCC1=CNC2=CC=CC=C21
C12H14N2O
NVUGEQAEQJTCIX-UHFFFAOYSA-N
70547
N-[2-(1H-indol-3-yl)ethyl]acetamide
Specifications
50mg
1016-47-3
n-acetyltryptamine, n-2-1h-indol-3-yl ethyl acetamide, 3-2-n-acetylaminoethyl indole, acetamide, n-2-1h-indol-3-yl ethyl, chembl33171, n-2-1h-indol-3-yl-ethyl-acetamide, acetotryptamide, acetamide, n-2-indol-3-ylethyl
CC(=O)NCCC1=CNC2=CC=CC=C21
202.257
CHEBI:55515
>99%
N-Acetyltryptamine
C12H14N2O
NVUGEQAEQJTCIX-UHFFFAOYSA-N
N-[2-(1H-indol-3-yl)ethyl]acetamide
70547
202.26