missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Acetylthiourea, 98%

Brand:  Alfa Aesar™ 047008.14

Code : NEW

Additional Details : CAS Number : 591-08-2 Weight : 0.02500kg Transport : UN number : 2811 Chem class : 1 Pack group : II

Quantity 25g

 View more versions of this product

Product Code. 15412827

  • £11.60 / 25g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Acetylthiourea
C3H6N2OS
MFCD00004937
969960
591-08-2
118.16
UN2811

Specifications

N-Acetylthiourea
Pale cream
25g
Crystalline Powder
C3H6N2OS
MFCD00004937
969960
Odorless
165° to 169°C
591-08-2
98%
118.16
UN2811
Videos
SDS