Learn More
N-Acetylglycyl-D-glutamic acid, Tocris Bioscience™
Potent convulsant
Brand:  Tocris (Bio-Techne) 039510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13467613
  • £73.50 / 10mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Excitatory peptide, more potent than L-glutamic acid at inducing seizures in mice.
Specifications
Chemical Identifiers
135701-69-8
246.219
n-acetylglycyl-d-glutamic acid, 2r-2-2-acetamidoacetamido pentanedioic acid, tocris-0395, r-2-2-acetamidoacetamido pentanedioic acid, 2r-2-2-acetamidoacetyl amino pentanedioic acid
(2R)-2-[(2-acetamidoacetyl)amino]pentanedioic acid
C9H14N2O6
CQTYJLQESPLSOP-ZCFIWIBFSA-N
6604718
CC(=O)NCC(=O)NC(CCC(=O)O)C(=O)O
Specifications
N-Acetylglycyl-D-glutamic acid
C9H14N2O6
CQTYJLQESPLSOP-ZCFIWIBFSA-N
(2R)-2-[(2-acetamidoacetyl)amino]pentanedioic acid
6604718
>98%
135701-69-8
n-acetylglycyl-d-glutamic acid, 2r-2-2-acetamidoacetamido pentanedioic acid, tocris-0395, r-2-2-acetamidoacetamido pentanedioic acid, 2r-2-2-acetamidoacetyl amino pentanedioic acid
CC(=O)NCC(=O)NC(CCC(=O)O)C(=O)O
246.219
246.22