Learn More
Alfa Aesar™ N-Acetyl-Val-Asp-Val-Ala-Asp-p-nitroanilide
Chromogenic substrate for caspase-2
Brand:  Alfa Aesar™ J65299.MB
Code : NEW
Additional Details : Weight : 0.00025kg
 View more versions of this product
Product Code. 15476819
  • £208.40 / 25mg
Estimated Shipment date
from Supplier 07-02-2018
Add to basket
Specifications
Specifications
25mg
−30° to −10°C
Chromogenic substrate for caspase-2
679.67g/mol
Signal Transduction Reagent
White to Off-white
Lyophilized Powder
C29H41N7O12
Ac-VDVAD-pNA; Caspase-2 substrate (chromogenic)