missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-Val-Asp-Val-Ala-Asp-7-amino-4-methylcoumarin, Thermo Scientific™
£139.00 - £434.00
Specifications
Description | White to Off-white |
---|---|
Content And Storage | -30°C to -10°C |
Molecular Formula | C33H44N6O12 |
Formula Weight | 716.73g/mol |
Synonym | Ac-VDVAD-AMC; Caspase-2 Substrate (ICH-1) fluorogenic |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15480497
|
Thermo Scientific Alfa Aesar
J66089.#0 |
5 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15490497
|
Thermo Scientific Alfa Aesar
J66089.MB |
25 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White to Off-white | |
C33H44N6O12 | |
Ac-VDVAD-AMC; Caspase-2 Substrate (ICH-1) fluorogenic | |
Lyophilized Powder |
-30°C to -10°C | |
716.73g/mol | |
Signal Transduction Reagent |