missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-S-(N-methylcarbamoyl)-L-cysteine, TRC

Product Code. 30010367
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010367 100 mg 100mg
17983175 10 mg 10mg
17920675 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010367 Supplier Toronto Research Chemicals Supplier No. TRCA186625100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103974-29-4
Molecular Formula C7 H12 N2 O4 S
Molecular Weight (g/mol) 220.25
Synonym L-Cysteine, N-acetyl-S-[(methylamino)carbonyl]- (ACI), L-Cysteine, N-acetyl-, methylcarbamate (ester) (9CI), N-Acetyl-S-[(methylamino)carbonyl]-L-cysteine (ACI), (2R)-2-Acetamido-3-[(methylcarbamoyl)sulfanyl]propanoic acid, (R)-2-Acetamido-3-((methylcarbamoyl)thio)propanoic acid, N-Acetyl-S-(N-methylcarbamoyl)cysteine, N-Acetyl-S-(N-methylcarbamoyl)-L-cysteine
IUPAC Name 2-acetamido-3-(methylcarbamoylsulfanyl)propanoic acid
SMILES CNC(=O)SCC(NC(=O)C)C(=O)O

Specifications

InChI Formula InChI=1S/C7H12N2O4S/c1-4(10)9-5(6(11)12)3-14-7(13)8-2/h5H,3H2,1-2H3,(H,8,13)(H,9,10)(H,11,12)
Quantity 100 mg
Formula Weight 220.0518
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-Acetyl-S-(N-methylcarbamoyl)-L-cysteine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.