Learn More
Tocris Bioscience™ N-Acetyl-N-acetoxy-4-chlorobenzenesulfonamide
Brand:  Tocris Bioscience™ 072210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13467893
  • £154.35 / 10mg
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
A pro-drug of the potent vasorelaxant nitroxyl, H-N=O, which it slowly releases in neutral solution. HNO eventually dimerizes and dehydrates to N2O, without producing NO. Also a powerful inhibitor of aldehyde dehydrogenase.
Specifications
Specifications
142867-52-5
Observed MW: 291.71
Soluble to 100mM in DMSO
C10H10ClNO5S