missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-tyrosine ethyl ester monohydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-3777150
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
36546-50-6
C13H19NO5
269.30
MFCD00149093
YWAVLHZJMWEYTA-HVPILOLGNA-N
n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
2723594
ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O
This item is not returnable. View return policy
36546-50-6
C13H19NO5
269.30
MFCD00149093
YWAVLHZJMWEYTA-HVPILOLGNA-N
n-acetyl-l-tyrosine ethyl ester monohydrate, ethyl n-acetyl-l-tyrosinate hydrate, ac-tyr-oet.h2o, atee, ethyl 2s-2-acetamido-3-4-hydroxyphenyl propanoate hydrate, n-acetyl-l-tyrosineethylestermonohydrate, n-acetyltyrosine ethyl ester monohydrate, ksc491m4n, n-acetyl tyrosine ethyl ester mono hydrate, acetyl-l-tyrosine ethyl ester hydrate
2723594
ethyl (2S)-2-acetamido-3-(4-hydroxyphenyl)propanoate;hydrate
O.CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 80°C to 81°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula 4-(HO)C6H4CH2CH(NHCOCH3)CO2C2H5·H2O
Quantity 1 g
Specific Rotation + 23
Solubility Information Solubility in water: soluble in cold water. Other solubilities: soluble in hot water,soluble in ethanol
Formula Weight 269.29
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-L-tyrosine ethyl ester monohydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'