missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-l-proline, 99%, Thermo Scientific Chemicals

Catalog Number p-3212933
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
68-95-1
C7H11NO3
157.17
MFCD00020837
GNMSLDIYJOSUSW-UHFFFAOYNA-N
n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid
66141
CHEBI:21560
(2S)-1-acetylpyrrolidine-2-carboxylic acid
CC(=O)N1CCCC1C(O)=O
This item is not returnable. View return policy
68-95-1
C7H11NO3
157.17
MFCD00020837
GNMSLDIYJOSUSW-UHFFFAOYNA-N
n-acetyl-l-proline, acetylproline, ac-pro-oh, n-acetylproline, 1-acetyl-l-proline, acetyl proline, l-proline, 1-acetyl, acetyl-l-proline, 2s-1-acetylpyrrolidine-2-carboxylic acid, s-1-acetylpyrrolidine-2-carboxylic acid
66141
CHEBI:21560
(2S)-1-acetylpyrrolidine-2-carboxylic acid
CC(=O)N1CCCC1C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 115°C to 117°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −86° (20°C c=1 EtOH)
Specific Rotation -86
Formula Weight 157.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Acetyl-L-proline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'