Learn More
N-Acetyl-L-leucyl-L-leucyl-L-methional, Tocris Bioscience™
Cathepsin inhibitor
Brand:  Tocris (Bio-Techne) 038410
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13457603
  • £147.00 / 10mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Very potent inhibitor of cathepsin L (Ki = 0.6 nM) and the strongest inhibitor of cathepsin B (Ki = 100 nM) amongst the peptide aldehydes. Also inhibits processing of malaria aspartic hemoglobinases plasmepsins I and II in vitro.
Specifications
Specifications
N-Acetyl-L-leucyl-L-leucyl-L-methional
136632-32-1
allm calpain inhibitor, calpain inhibitor ii, n-acetyl-l-leucyl-l-leucyl-l-methioninal, calpain inhibitor ii, allm, l-leucinamide, n-acetyl-l-leucyl-n-1-formyl-3-mercaptopropyl-, s
CC(C)CC(C(=O)NC(CC(C)C)C(=O)NC(CCSC)C=O)NC(=O)C
401.566
401.56
10mg
C19H35N3O4S
RJWLAIMXRBDUMH-CGZBRXJRSA-N
(2S)-2-acetamido-4-methyl-N-[4-methyl-1-[(4-methylsulfanyl-1-oxobutan-2-yl)amino]-1-oxopentan-2-yl]pentanamide
16218939
>98%