missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-glutamine, 97%, Thermo Scientific™

Catalog Number p-4813568
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
35305-74-9
C7H12N2O4
188.18
MFCD00038159
KSMRODHGGIIXDV-YFKPBYRVSA-N
n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
182230
CHEBI:21553
CC(=O)N[C@@H](CCC(N)=O)C(O)=O
This item is not returnable. View return policy
35305-74-9
C7H12N2O4
188.18
MFCD00038159
KSMRODHGGIIXDV-YFKPBYRVSA-N
n-acetyl-l-glutamine, aceglutamide, l-glutamine, n-acetyl, nalpha-acetyl-l-glutamine, s-2-acetamido-5-amino-5-oxopentanoic acid, acetylglutamine, n-alpha-acetyl-l-glutamine, n-acetylglutamine, acutil-s, acetyl-l-glutamine
182230
CHEBI:21553
CC(=O)N[C@@H](CCC(N)=O)C(O)=O
Melting Point 206°C
Ignition Residue 0.2% max
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
IUPAC Name (2S)-4-carbamoyl-2-acetamidobutanoic acid
Formula Weight 188.18
Percent Purity 97%
Chemical Name or Material N-Acetyl-L-glutamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'