missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-L-glutamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Specifications
Melting Point | 206°C |
Ignition Residue | 0.2% max |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Quantity | 25 g |
Solubility Information | Solubility in water: soluble |
IUPAC Name | (2S)-2-acetamido-5-amino-5-oxopentanoic acid |
Formula Weight | 188.18 |
Percent Purity | 97% |
Chemical Name or Material | N-Acetyl-L-glutamine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'