missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-cysteine, Merck Millipore

For biochemistry

Brand:  Merck Millipore 1124220100

Code : NEW

Additional Details : CAS Number : 616-91-1 Weight : 0.10000kg

Quantity 100 g

 View more versions of this product

Product Code. 10026310

Quantity Price
1 £104.00 / 100g
10 £89.40 / 100g
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

  • Assay (iodometric): ≥ 99.0 %
  • Identity (IR-spectrum): passes test
  • Appearance: white to almost white, crystalline powder or colorless crystals
  • Appearance of solution (1 mol/L, water): clear and colorless
  • Spec. rotation (α 20/D, 50 g/L, buffer pH 7.0): +21.0 to +27.0
  • Heavy metals (as Pb): ≤ 0.001 %
  • NH4 (Ammonium): ≤ 0.01 %
  • L-cysteine (HPLC): ≤ 0.02 %
  • Other ninhydrine positive substances: ≤ 0.1 %
Specifications

Chemical Identifiers

616-91-1
1.48 g/cm3 (at 20°C)
PWKSKIMOESPYIA-UHFFFAOYNA-N
MFCD00004880
1.5-2.5 (20°C, 100 g/L in H2O)
2-Acetamido-3-mercaptopropionic acid
5050 mg/kg LD50, oral (Rat)
1 S/C5H9NO3S/c1-3(7)6-4(2-10)5(8)9/h4,10 H,2H2,1H3,(H,6,7)(H,8,9)
C5H9NO3S
163.19
179.5 g/L

Specifications

616-91-1
C5H9NO3S
2-Acetamido-3-mercaptopropionic acid
PWKSKIMOESPYIA-UHFFFAOYNA-N
2-acetamido-3-sulfanylpropanoic acid
179.5 g/L
1.5-2.5 (20°C, 100 g/L in H2O)
5050 mg/kg LD50, oral (Rat)
C5H9NO3S
MFCD00004880
1 S/C5H9NO3S/c1-3(7)6-4(2-10)5(8)9/h4,10 H,2H2,1H3,(H,6,7)(H,8,9)
CC(=O)NC(CS)C(O)=O
163.19
1.48 g/cm3 (at 20°C)
100 g
Videos
SDS
Documents
Certificates