missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-L-alanine methyl ester, 95%, Thermo Scientific™

Catalog Number p-7057375
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3619-02-1
MFCD03940666
This item is not returnable. View return policy
3619-02-1
MFCD03940666

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Yellow
Molecular Formula C6H11NO3
Beilstein 1723226
Synonym s-methyl 2-acetamidopropanoate, ac-ala-ome, n-acetyl-l-alanine methyl ester, methyl 2s-2-acetamidopropanoate, l-alanine, n-acetyl-, methyl ester, alanine, n-acetyl-,methyl ester, ac-l-ala-ome, ambotzaaa1912, n-acetyl-l-alanine methyl, n-acetylalanine methyl ester
InChI Key FQGVVDYNRHNTCK-UHFFFAOYNA-N
SMILES CC(C(=O)OC)NC(=O)C
IUPAC Name methyl (2S)-2-acetamidopropanoate
Molecular Weight (g/mol) 145.16
PubChem CID 6951030
Formula Weight 145.16
Percent Purity 95%
Quantity 5 g
Physical Form Liquid
Chemical Name or Material N-Acetyl-L-alanine methyl ester
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'