missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Acetyl-D-sphingosine, 98%

Catalog Number p-7082236
Click to view available options
Quantity:
1 mg
5 mg
10 mg
This item is not returnable. View return policy
3102-57-6
C20H39NO3, C20H39NO3
Acetyl ceramide; C2-Ceramide
This item is not returnable. View return policy
3102-57-6
C20H39NO3, C20H39NO3
Acetyl ceramide; C2-Ceramide

Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells
  • Activates protein phosphatases
TRUSTED_SUSTAINABILITY
Quantity 1 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Inhibits cell proliferation and induces monocytic differentiation of HL-60 cells. Activates protein phosphatases
Formula Weight 341.53g/mol
Melting Point 77°C to 78°C
Solubility Soluble in ethanol, methanol, chloroform or DMSO at approximately 10mg/ml
Product Type Signal Transduction Reagent
Form Powder
InChI Key BLTCBVOJNNKFKC-QUDYQQOWSA-N
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NC(C)=O
IUPAC Name N-[(2S,3R,4E)-1,3-dihydroxyoctadec-4-en-2-yl]acetamide
Molecular Weight (g/mol) 341.54
Assay 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'