missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Acetyl-D-phenylalanine, 95%

Brand:  Alfa Aesar™ H63382.14

Code : NEW

Additional Details : CAS Number : 10172-89-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15440049

  • £98.40 / 25g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

N-Acetyl-D-phenylalanine
207.23
2213852
10172-89-1
MFCD00002664
Ac-D-Phe-OH

Specifications

N-Acetyl-D-phenylalanine
Solid
207.23
2213852
White
25g
10172-89-1
95%
MFCD00002664
Ac-D-Phe-OH
165° to 169°C
Videos
SDS