missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-galactosamine, 98%, Thermo Scientific Chemicals

Catalog Number p-4228287
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1811-31-0
C8H15NO6
221.21
MFCD00136044
OVRNDRQMDRJTHS-KEWYIRBNSA-N
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
60196345
N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O
This item is not returnable. View return policy
1811-31-0
C8H15NO6
221.21
MFCD00136044
OVRNDRQMDRJTHS-KEWYIRBNSA-N
dsstox_cid_28637, dsstox_rid_82907, dsstox_gsid_48711, n-acetyl-, a-d-galactosamine
60196345
N-[(2R,3S,4S,5S)-3,4,5,6-tetrahydroxy-1-oxohexan-2-yl]acetamide
CC(=O)N[C@H]1C(O)O[C@H](CO)[C@H](O)[C@@H]1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 158.0°C to 166.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation 85.3
Solubility Information (10% in water) Clear colorless
Formula Weight 221.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-D-galactosamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'