missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Acetyl-3,5-dinitro-L-tyrosine, 98%

Brand:  Alfa Aesar™ H63186.06

Code : NEW

Additional Details : CAS Number : 20767-00-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15479738

  • £208.82 / 5g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

N-Acetyl-3,5-dinitro-L-tyrosine
313.22
Ac-tyr(3,5-NO2)-OH
20767-00-4
MFCD00024239

Specifications

N-Acetyl-3,5-dinitro-L-tyrosine
Solid
313.22
Ac-tyr(3,5-NO2)-OH
20767-00-4
98%
MFCD00024239
5g
Videos
SDS