missing translation for 'onlineSavingsMsg'
Learn More

N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Catalog Number 10606544
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
82911-69-1
C19H15NO5
337.29
MFCD00010733
WMSUFWLPZLCIHP-UHFFFAOYSA-N
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide
134122
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
This item is not returnable. View return policy
82911-69-1
C19H15NO5
337.29
MFCD00010733
WMSUFWLPZLCIHP-UHFFFAOYSA-N
fmoc-osu, n-9-fluorenylmethoxycarbonyloxy succinimide, fmoc-onsu, 9-fluorenylmethyl succinimidyl carbonate, fmoc n-hydroxysuccinimide ester, 9h-fluoren-9-yl methyl 2,5-dioxopyrrolidin-1-yl carbonate, 9-fluorenylmethyl n-succinimidyl carbonate, 9-fluorenylmethylsuccinimidyl carbonate, 9-fluorenylmethoxycarbonyl-n-hydroxysuccinimide
134122
(2,5-dioxopyrrolidin-1-yl) 9H-fluoren-9-ylmethyl carbonate
C1CC(=O)N(C1=O)OC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 145.0°C to 153.0°C
Color White
Loss on Drying 0.25% max. (60°C, 2 hrs, 1mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 337.29
Percent Purity 98%
Physical Form Microfine Powder
Chemical Name or Material N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • N-(9H-Fluoren-2-ylmethoxycarbonyloxy)succinimide
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'