missing translation for 'onlineSavingsMsg'
Learn More

N-(6-Bromohexyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog Number p-7028018
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
24566-79-8
C14H16BrNO2
310.191
MFCD00023098
OAZFTIPKNPTDIO-UHFFFAOYSA-N
141120
2-(6-bromohexyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr
This item is not returnable. View return policy
24566-79-8
C14H16BrNO2
310.191
MFCD00023098
OAZFTIPKNPTDIO-UHFFFAOYSA-N
141120
2-(6-bromohexyl)isoindole-1,3-dione
C1=CC=C2C(=C1)C(=O)N(C2=O)CCCCCCBr

N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(6-Bromohexyl)phthalimide is used as a pharmaceutical intermediate and in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 54°C to 60°C
Boiling Point 186°C to 188°C (0.4 mmHg)
Odor Odorless
Quantity 5 g
Beilstein 186875
Synonym n-6-bromohexyl phthalimide, 2-6-bromohexyl isoindoline-1,3-dione, n-6-bromohexyl-phthalimide, 2-6-bromohexyl-1h-isoindole-1,3 2h-dione, 2-6-bromohexyl isoindole-1,3-dione, 2-6-bromohexyl-2,3-dihydro-1h-isoindole-1,3-dione, 2-6-bromo-hexyl-isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-6-bromohexyl, 1h-isoindole-1,3 2h-dione, 2-6-bromohexyl, 6-n-phthalimido hexyl bromide
Solubility Information Insoluble in water.
Formula Weight 310.19
Percent Purity 97%
Chemical Name or Material N-(6-Bromohexyl)phthalimide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'