missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Hexynyl)phthalimide, 97%, Thermo Scientific Chemicals

Catalog Number p-7035762
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
6097-08-1
MFCD00671372
This item is not returnable. View return policy
6097-08-1
MFCD00671372

6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-amino-1-hexyne was prepared from N-(5-Hexynyl)phthalimide which is the starting material. Used in the synthesis of Amino-alkyne.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 68°C to 72°C
Molecular Formula C10H10O3
Quantity 5 g
Synonym 3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid
InChI Key LZPNXAULYJPXEH-AATRIKPKSA-N
SMILES COC1=CC=CC(=C1)C=CC(=O)O
IUPAC Name (E)-3-(3-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 178.19
PubChem CID 637668
Formula Weight 227.26
Percent Purity 97%
Chemical Name or Material N-(5-Hexynyl)phthalimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'