missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(5-Chloro-2-methylphenyl)-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59239.03

Code : NEW

Additional Details : CAS Number : 304890-11-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15445488

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(5-Chloro-2-methylphenyl)-3-methylbenzamide
304890-11-7
259.74
MFCD00684371

Specifications

N-(5-Chloro-2-methylphenyl)-3-methylbenzamide
Solid
259.74
1g
304890-11-7
97%
MFCD00684371
Videos
SDS