missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(5,6,7,8-Tetrahydro-1-naphthyl)-3,5-bis(trifluoromethyl)benzamide, 95%

Brand:  Alfa Aesar™ H65897.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409986

  • £74.73 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

N-(5,6,7,8-Tetrahydro-1-naphthyl)-3,5-bis(trifluoromethyl)benzamide
387.33
MFCD11224011

Specifications

N-(5,6,7,8-Tetrahydro-1-naphthyl)-3,5-bis(trifluoromethyl)benzamide
Solid
387.33
1g
95%
MFCD11224011
Videos
SDS