missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-(5,6,7,8-Tetrahydro-1-naphthyl)-2,5-bis(trifluoromethyl)benzamide, 95%
Brand: Alfa Aesar™ H65210.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N-(5,6,7,8-Tetrahydro-1-naphthyl)-2,5-bis(trifluoromethyl)benzamide | |
387.33 | |
MFCD27975734 |
Specifications
N-(5,6,7,8-Tetrahydro-1-naphthyl)-2,5-bis(trifluoromethyl)benzamide | |
Solid | |
387.33 |
1g | |
95% | |
MFCD27975734 |