missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar N-[4-(Trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar H60648.03

N-[4-(Trifluoromethyl)phenyl]benzenesulfonamide, 97%

Code : NEW

Additional Details : CAS Number : 433-03-4 Weight : 0.00100kg

Product Code. 15485916

  • £358.49 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket