missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-[4-(Trifluoromethyl)phenyl]-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65026.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429326

  • £66.46 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

N-[4-(Trifluoromethyl)phenyl]-5,6,7,8-tetrahydronaphthalene-1-carboxamide
319.33
MFCD28098737

Specifications

N-[4-(Trifluoromethyl)phenyl]-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
319.33
1g
95%
MFCD28098737
Videos
SDS