missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nalpha-4-Tosyl-L-arginine methyl ester hydrochloride, 98+%

Catalog Number 10755981
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1784-03-8
C14H22N4O4S·HCl
378.88
MFCD00012578
JIQFFACVQXXHMY-YDALLXLXSA-N
tos-arg-ome.hcl, tame hydrochloride, tos-arg-ome hcl, nalpha-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride, tos-arg-omehcl, s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride, tosyl-l-arginine methyl ester hydrochloride, n-4-tosyl-l-arginine methyl ester hydrochloride
2723792
methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl
This item is not returnable. View return policy
1784-03-8
C14H22N4O4S·HCl
378.88
MFCD00012578
JIQFFACVQXXHMY-YDALLXLXSA-N
tos-arg-ome.hcl, tame hydrochloride, tos-arg-ome hcl, nalpha-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-tosyl-l-arginine methyl ester hydrochloride, nalpha-p-toluenesulfonyl-l-arginine methyl ester hydrochloride, tos-arg-omehcl, s-methyl 5-guanidino-2-4-methylphenylsulfonamido pentanoate hydrochloride, tosyl-l-arginine methyl ester hydrochloride, n-4-tosyl-l-arginine methyl ester hydrochloride
2723792
methyl (2S)-5-(diaminomethylideneamino)-2-[(4-methylphenyl)sulfonylamino]pentanoate;hydrochloride
CC1=CC=C(C=C1)S(=O)(=O)NC(CCCN=C(N)N)C(=O)OC.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 146°C to 149°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)3CH(NHSO2C6H4CH3)CO2CH3
Quantity 10 g
Specific Rotation -14
Formula Weight 378.88
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Nα-4-Tosyl-L-arginine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'