missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-tert-Butylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60998.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15447488

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-tert-Butylphenyl)benzenesulfonamide
289.39
MFCD07957302

Specifications

N-(4-tert-Butylphenyl)benzenesulfonamide
1g
97%
MFCD07957302
Pink
Solid
289.39
Videos
SDS