missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-tert-Butylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65835.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462639

  • £99.64 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-tert-Butylphenyl)-2,2-diphenylacetamide
343.47
MFCD04678678

Specifications

N-(4-tert-Butylphenyl)-2,2-diphenylacetamide
Solid
343.47
1g
95%
MFCD04678678
Videos
SDS