missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Oxocyclohexyl)acetamide, 98%

Brand:  Alfa Aesar™ H64511.14

Code : NEW

Additional Details : CAS Number : 27514-08-5 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15421759

  • £46.30 / 25g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Oxocyclohexyl)acetamide
155.2
2803083
27514-08-5
MFCD03703462
4-Acetamidocyclohexanone

Specifications

N-(4-Oxocyclohexyl)acetamide
136° to 140°C
27514-08-5
98%
MFCD03703462
4-Acetamidocyclohexanone
Off-white
25g
Solid
155.2
2803083
Videos
SDS