missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-n-Butylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65994.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15480077

  • £70.28 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-n-Butylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
307.44
MFCD28100497

Specifications

N-(4-n-Butylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
307.44
1g
95%
MFCD28100497
Videos
SDS