missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-n-Butylphenyl)-4-chloro-2-fluoro-5-sulfamoylbenzamide, 95%

Brand:  Alfa Aesar™ H65059.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499346

  • £119.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-n-Butylphenyl)-4-chloro-2-fluoro-5-sulfamoylbenzamide
384.85
MFCD28100495

Specifications

N-(4-n-Butylphenyl)-4-chloro-2-fluoro-5-sulfamoylbenzamide
Solid
384.85
1g
95%
MFCD28100495
Videos
SDS