missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-n-Butylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65470.03

Code : NEW

Additional Details : CAS Number : 5404-36-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15492559

  • £91.90 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-n-Butylphenyl)-2,2-diphenylacetamide
5404-36-4
343.47
MFCD00686863

Specifications

N-(4-n-Butylphenyl)-2,2-diphenylacetamide
5404-36-4
95%
MFCD00686863
1g
Solid
343.47
Videos
SDS