missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60666.03

Code : NEW

Additional Details : CAS Number : 6311-65-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15427148

  • £358.49 / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Methylphenyl)benzenesulfonamide
6311-65-5
247.31
MFCD00594268

Specifications

N-(4-Methylphenyl)benzenesulfonamide
6311-65-5
97%
MFCD00594268
1g
Solid
247.31
Videos
SDS