missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide, 95%

Brand:  Alfa Aesar™ H65411.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449626

  • £119.78 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
265.36
MFCD28098793
N-(p-Tolyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide

Specifications

N-(4-Methylphenyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
Solid
265.36
N-(p-Tolyl)-5,6,7,8-tetrahydronaphthalene-1-carboxamide
1g
95%
MFCD28098793
Videos
SDS