missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-(4-Methylphenyl)-2,2-diphenylacetamide, 95%

Brand:  Alfa Aesar™ H65712.03

Code : NEW

Additional Details : CAS Number : 4107-01-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15482609

  • £127.20 / 1g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

N-(4-Methylphenyl)-2,2-diphenylacetamide
301.39
2,2-Diphenyl-N-p-tolylacetamide
4107-01-1
MFCD00228877

Specifications

N-(4-Methylphenyl)-2,2-diphenylacetamide
4107-01-1
95%
MFCD00228877
1g
Solid
301.39
2,2-Diphenyl-N-p-tolylacetamide
Videos
SDS